missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bromide, ultra dry, 99.98% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Potassium bromide is used in manufacturing of photographic papers, engraving and lithography, in spectroscopy and as an analytical lab reagent. Potassium bromide is employed in antiepileptic medication for dogs. It is utilized in the bromination of alkenes using cerium(IV) ammonium nitrate. It plays an essential role with diacetoxy iodobenzene for bromination of alkenes and alkynes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Boiling Point | 1435°C |
Melting Point | 730°C |
Color | White |
Physical Form | Beads |
Quantity | 25 g |
Merck Index | 14,7618 |
Solubility Information | Freely soluble in water. Soluble in alcohol and glycerol. Slightly soluble in diethyl ether. |
IUPAC Name | potassium;bromide |
Formula Weight | 119.01 |
Percent Purity | 99.98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'