missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, Free-flowing, ACS reagent, anhydrous, ≥99%, Honeywell Fluka™

Catalog Number 15637000
Click to view available options
Quantity:
100 g
500 g
1 kg
6 x 500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
potassium;bromide
[K+].[Br-]
This item is not returnable. View return policy
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
potassium;bromide
[K+].[Br-]

Free-flowing, ACS reagent, anhydrous,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Boiling Point 1,383°C
Melting Point 734°C (literature)
CAS Max % 100.0000%
Physical Form Anhydrous
Quantity 1 kg
Linear Formula KBr
Formula Weight 119
Percent Purity >99%
Grade ACS Reagent
Packaging Poly Bottle
Density 2.73 g/cm3
Chemical Name or Material Potassium bromide, Free-flowing, ACS reagent, anhydrous, ≥99%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'