missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical P/3960/60

Code : 21

Additional Details : CAS Number : 7758-02-3 Weight : 1.00000kg


Quantity 1kg

 View more versions of this product

Product Code. 10131770

  • £85.00 / 1kg

In stock, ready to ship: 16

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potassium Bromide, Extra Pure, SLR

Specifications

Chemical Identifiers

7758-02-3
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
253877
potassium;bromide
BrK
11358
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
CHEBI:32030
[K+].[Br-]

Specifications

7758-02-3
11358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium;bromide
253877
119g/mol
Plastic powder jar
1435°C
734°C
1kg
BrK
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
[K+].[Br-]
119.00
CHEBI:32030
Solid
<0.01mmHg at 20°C
White
5 to 8.8
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium bromide

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS Documents
Certificates