missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium bromide crystal optic rectangle, 41mm x 23mm x 6mm, unpolished

Brand:  Alfa Aesar™ 038411.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15406215

  • £37.90 / Each


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Specifications

Potassium bromide crystal optic rectangle
41mm x 23mm x 6mm, Unpolished
1 each
Videos
SDS