missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bromide crystal optic rectangle, 41mm x 23mm x 6mm (drilled), polished both sides, Thermo Scientific™
Click to view available options
Quantity:
1 Ea.
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS | 7758-02-3 |
Quantity | 1 Ea. |
Molecular Formula | BrK |
MDL Number | MFCD00011358 |
Synonym | potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 |
InChI Key | IOLCXVTUBQKXJR-UHFFFAOYSA-M |
SMILES | [K+].[Br-] |
IUPAC Name | potassium bromide |
Molecular Weight (g/mol) | 119.00 |
PubChem CID | 253877 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'