missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium bromide crystal optic rectangle, 38.5mm x 19.5mm x 4mm, unpolished

Brand:  Alfa Aesar™ 038409.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15496205

  • £19.00 / Each


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Specifications

Potassium bromide crystal optic rectangle
38.5mm x 19.5mm x 4mm, Unpolished
7758-02-3
1 each
Videos
SDS