missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium bromide crystal optic disc, 25mm x 2mm, polished both sides

Brand:  Alfa Aesar™ 038451.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15436245

  • £36.20 / Each


Estimated Shipment date
from Supplier 25-04-2018
Add to basket

Specifications

Specifications

Potassium bromide crystal optic disc
25mm x 2mm, Polished both sides
MFCD00011358
1 each
Videos
SDS