missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium bromide crystal optic disc, 13mm x 2mm, unpolished

Spectroscopy, IR window

Brand:  Alfa Aesar™ 038391.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15496195

  • £14.90 / Each


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Specifications

Potassium bromide crystal optic disc
13mm x 2mm, Unpolished
MFCD00011358
1 each
Videos
SDS