missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, BAKER INSTRA-ANALYZED™ Reagent, J.T. Baker™

Catalog Number p-5390812
1173.25 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
100 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
[K+].[Br-]
This item is not returnable. View return policy
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
[K+].[Br-]

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

    For Infrared Analysis
Quantity 100 g
IUPAC Name potassium;bromide
Formula Weight 118.99
Grade BAKER INSTRA-ANALYZED™ Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.75kg
Chemical Name or Material Potassium Bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'