missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bromide, 99+%, for analysis, Thermo Scientific Chemicals
14.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Specifications
Boiling Point | 1435.0°C |
Melting Point | 734.0°C |
Color | White |
pH | 5.0 to 8.8 (5% soln. at 20°C) |
Physical Form | Crystalline Powder, Crystals and/or Chunks |
Quantity | 100 g |
Assay Percent Range | 99% min. (Argentometry) |
Linear Formula | KBr |
Merck Index | 15, 7739 |
Solubility Information | Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C), 1.42g/L ethanol (25°C), 217g/L glycerol, 4g/L alcohol |
Show More |
Safety and Handling
Product Identifier
- Potassium bromide
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'