missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bromate/Potassium bromide, 0.1N Standardized Solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
4 L
Description
Potassium bromate/Potassium bromide is used as an analytical reagent for volumetric analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 1 L |
Color | Colorless |
Odor | Odorless |
Solubility | Miscible with water |
Physical Form | Liquid |
Chemical Name or Material | Potassium bromate/Potassium bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'