missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bromate, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 350.0°C |
Quantity | 500 g |
Linear Formula | KBrO3 |
Merck Index | 15,7738 |
Solubility Information | Solubility in water: 70g/L (20°C). Other solubilities: practically insoluble in alcohol, soluble in boiling water |
Formula Weight | 167.01 |
Grade | ACS Reagent |
Packaging | Plastic bottle |
Specific Gravity | 3.27 |
Density | 3.2700g/mL |
Show More |
Safety and Handling
Product Identifier
- Potassium bromate
- Danger
- Oxidising solid Category 1
- Acute toxicity Category 3
- Carcinogenicity Category 1B
- H271-May cause fire or explosion; strong oxidiser.
- H301-Toxic if swallowed.
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- REA17-CMR1-Restricted to professional users.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'