missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromate, ACS, ≥99.8% (Dried material, RT), Honeywell Fluka™

Catalog Number p-7120682
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly bottle
7758-01-2
BrKO3
167.00
MFCD00011359
OCATYIAKPYKMPG-UHFFFAOYSA-M
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
23673461
CHEBI:38211
potassium;bromate
[O-]Br(=O)=O.[K+]
7758-01-2
BrKO3
167.00
MFCD00011359
OCATYIAKPYKMPG-UHFFFAOYSA-M
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
23673461
CHEBI:38211
potassium;bromate
[O-]Br(=O)=O.[K+]

Puriss. p.a., ACS Reagent,≥99.8% (dried material, RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Color Colorless or White
pH 5.0 to 9.0 (at 25°C, 50mg/mL in H2O)
Quantity 50 g
Linear Formula KBrO3
UN Number UN1484
Formula Weight 167g/mol
Percent Purity ≥99.8% (dried material, RT)
Packaging Poly bottle
Density 3.270g/mL
Chemical Name or Material Potassium bromate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium bromate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Oxidising solid Category 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H301-Toxic if swallowed.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : potassium bromate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'