missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromate, ACS, ≥99.8% (Dried material, RT), Honeywell Fluka™

Catalog Number 15635910
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly bottle
This item is not returnable. View return policy
7758-01-2
BrKO3
167.00
MFCD00011359
OCATYIAKPYKMPG-UHFFFAOYSA-M
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
23673461
CHEBI:38211
potassium;bromate
[O-]Br(=O)=O.[K+]
This item is not returnable. View return policy
7758-01-2
BrKO3
167.00
MFCD00011359
OCATYIAKPYKMPG-UHFFFAOYSA-M
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
23673461
CHEBI:38211
potassium;bromate
[O-]Br(=O)=O.[K+]

Puriss. p.a., ACS Reagent,≥99.8% (dried material, RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Color Colorless or White
pH 5.0 to 9.0 (at 25°C, 50mg/mL in H2O)
Quantity 250 g
Linear Formula KBrO3
UN Number UN1484
Formula Weight 167g/mol
Percent Purity ≥99.8% (dried material, RT)
Packaging Poly bottle
Density 3.270g/mL
Chemical Name or Material Potassium bromate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'