missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium borohydride, 98%

Brand:  Alfa Aesar™ 046847.A1

Code : NEW

Additional Details : CAS Number : 13762-51-1 Weight : 1.00000kg Transport : UN number : 1870 Chem class : 1 Pack group : I

Quantity 1kg

 View more versions of this product

Product Code. 15492777

  • £292.50 / 1kg


Estimated Shipment date
from Supplier 16-02-2018
Add to basket

Description

Description

  • Source of hydrogen, reducing agent for aldehydes, ketones, acid chlorides; foaming agent for plastics
Specifications

Chemical Identifiers

Potassium borohydride
BH4K
53.94
UN1870
Potassium tetrahydridoborate
13762-51-1
KBH4
MFCD00011396
14,7616

Specifications

Potassium borohydride
Granules
BH4K
53.94
UN1870
Potassium tetrahydridoborate
1.17g/mL
Odorless
Moisture Sensitive
1kg
13762-51-1
98%
KBH4
MFCD00011396
14,7616
Soluble in water (19g/100g ); methanol (3.9g/100g ); 4:1 water:methanol mixture (13g/100g );
−4 Mesh
1.494
>400°C (dec.)
Videos
SDS