missing translation for 'onlineSavingsMsg'
Learn More

Potassium bisulfate, 99%, extra pure, Thermo Scientific Chemicals

Catalog Number 13217069
21.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
7646-93-7
HKO4S
136.16
MFCD00011404
CHKVPAROMQMJNQ-UHFFFAOYSA-M
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
516920
[K+].OS([O-])(=O)=O
7646-93-7
HKO4S
136.16
MFCD00011404
CHKVPAROMQMJNQ-UHFFFAOYSA-M
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
516920
[K+].OS([O-])(=O)=O

Potassium bisulfate, 99%, HKO4S, CAS Number-7646-93-7, potassium bisulfate, sulfuric acid, monopotassium salt, sal enixum, potassium hydrogensulphate, potassium bisulphate, acid potassium sulfate, potassium acid sulfate, potassium hydrogen sulfate, monopotassium sulfate, caswell no. 682c, 500g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 214.0°C
Quantity 500 g
Assay Percent Range 99%
Linear Formula KHSO4
Merck Index 15,7734
Solubility Information (10% in water) Clear colorless
IUPAC Name potassium hydrogen sulfate
Formula Weight 136.16
Percent Purity 99%
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Potassium bisulfate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium bisulfate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'