missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium bis(trimethylsilyl)amide, 20% w/w, 0.91M in THF

Brand:  Alfa Aesar™ 035450.K2

Code : NEW

Additional Details : CAS Number : 40949-94-8 Weight : 1.00000kg Transport : UN number : 2924 Chem class : 3 Pack group : II

Quantity 1L

 View more versions of this product

Product Code. 15431347

  • £199.28 / 1 L


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

  • Used in the preparation of lanthanide complexes, selective cyclization reactions, and as an anionic initiator for polymerization
Specifications

Chemical Identifiers

Potassium bis(trimethylsilyl)amide
[(CH3)3Si]2NK
MFCD00010330
Potassium hexamethyldisilazide
40949-94-8
199.49
UN2924

Specifications

Potassium bis(trimethylsilyl)amide
Liquid
199.49
UN2924
0.910g/mL
Colorless to amber
1L
40949-94-8
[(CH3)3Si]2NK
MFCD00010330
Potassium hexamethyldisilazide
66°C
20% w/w, 0.91M in THF
Air & Moisture Sensitive
Videos
SDS