missing translation for 'onlineSavingsMsg'
Learn More

Potassium bis(trimethylsilyl)amide, 15% w/w in toluene, Alfa Aesar™

Brand:  Alfa Aesar 044143.AC

Code : NEW

Additional Details : CAS Number : 40949-94-8 Weight : 0.01000kg Transport : UN number : 2924 Chem class : 1 Pack group : II

Quantity 25mL

 View more versions of this product

Product Code. 11379638

  • £19.50 / 25mL


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

Potassium bis(trimethylsilyl)amide
Liquid
199.49
Potassium hexamethyldisilazide
40949-94-8
C6H18KNSi2
MFCD00010330

Specifications

Potassium bis(trimethylsilyl)amide
Liquid
[(CH3)3Si]2NK
MFCD00010330
4006754
Packaged under Argon in resealable ChemSeal™ bottles
0.881
15% w/w in toluene
7°C (44°F)
40949-94-8
C6H18KNSi2
199.49
UN2924
Potassium hexamethyldisilazide
1.4920
Colorless to Amber
25mL
Air and Moisture Sensitive
Videos
SDS