missing translation for 'onlineSavingsMsg'
Learn More

Potassium bis(trimethylsilyl)amide, 15% w/w in toluene, Alfa Aesar™

Brand:  Alfa Aesar 044143.AP

Code : NEW

Additional Details : CAS Number : 40949-94-8 Weight : 0.01000kg Transport : UN number : 2924 Chem class : 1 Pack group : II

Quantity 500mL

 View more versions of this product

Product Code. 11470680

  • £263.94 / 500mL


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Specifications

Chemical Identifiers

Potassium bis(trimethylsilyl)amide
Liquid
199.49
Potassium bis(trimethylsilyl)amide
40949-94-8
C6H18KNSi2
MFCD00010330

Specifications

Potassium bis(trimethylsilyl)amide
Liquid
199.49
Potassium bis(trimethylsilyl)amide
500mL
40949-94-8
C6H18KNSi2
MFCD00010330
0.881g/mL
Videos
SDS