missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Potassium bis(trifluoromethylsulfonyl)imide is used as Flavor & Fragrance intermediates. It is used as an intermediate in Industries such as agrochemical, dyestuff, pharmaceutical, syntheses material, chemical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 198°C to 203°C |
Linear Formula | (CF3SO2)2NK |
Quantity | 5 g |
UN Number | UN1759 |
Sensitivity | Moisture sensitive |
Solubility Information | It is soluble in water. 10mg/mL. |
Formula Weight | 319.24 |
Physical Form | Solid |
Chemical Name or Material | Potassium bis (trifluoromethylsulfonyl)imide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'