missing translation for 'onlineSavingsMsg'
Learn More

Potassium acetate hydrate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049126
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
254966-93-3
C2H3KO2
98.14
MFCD00012458
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate hydrate, c2h3o2.k.h2o, potassium acetate hydrate, puratronic, potassium acetate hydrate, puratronic r metals basis
23702041
[K+].CC([O-])=O
This item is not returnable. View return policy
254966-93-3
C2H3KO2
98.14
MFCD00012458
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate hydrate, c2h3o2.k.h2o, potassium acetate hydrate, puratronic, potassium acetate hydrate, puratronic r metals basis
23702041
[K+].CC([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline
Quantity 5 g
IUPAC Name potassium acetate
Formula Weight 98.15 (Anhydrous)
Percent Purity 99.997%
Grade Puratronic™
Assay (metals basis)
Chemical Name or Material Potassium acetate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'