missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acetate, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073402
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
127-08-2
C2H3KO2
98.142
12458
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
517044
CHEBI:32029
potassium;acetate
CC(=O)[O-].[K+]
This item is not returnable. View return policy
127-08-2
C2H3KO2
98.142
12458
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
517044
CHEBI:32029
potassium;acetate
CC(=O)[O-].[K+]

Potassium Acetate, Extra Pure, SLR

TRUSTED_SUSTAINABILITY
Melting Point 292°C
Color White
pH 7.0 to 8.0
Physical Form Solid
Quantity 500 g
Formula Weight 98.14g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'