missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acetate, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical P/3760/65

Code : 21

Additional Details : CAS Number : 127-08-2 Weight : 5.00000kg

Packaging Plastic keg
Quantity 5kg

 View more versions of this product

Product Code. 10080300

Quantity Price
1 £454.00 / 5kg
4 £408.56 / 5kg
16 £388.12 / 5kg
Estimated Shipment: 26-08-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Potassium Acetate, Extra Pure, SLR

Specifications

Chemical Identifiers

127-08-2
98.142
SCVFZCLFOSHCOH-UHFFFAOYSA-M
517044
potassium;acetate
C2H3KO2
12458
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
CHEBI:32029
CC(=O)[O-].[K+]

Specifications

127-08-2
12458
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium;acetate
517044
98.14g/mol
Plastic keg
292°C
5kg
C2H3KO2
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
CC(=O)[O-].[K+]
98.142
CHEBI:32029
Solid
White
7.0 to 8.0
Videos
SDS Documents
Certificates