missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium 4-acetylphenyltrifluoroborate, 97%

Brand:  Alfa Aesar™ H55845.14

Code : NEW

Additional Details : CAS Number : 252726-24-2 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15432218

  • £384.20 / 25g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

Potassium 4-acetylphenyltrifluoroborate
252726-24-2
226.05
MFCD07784376

Specifications

Potassium 4-acetylphenyltrifluoroborate
Powder
226.05
White to off-white
25g
252726-24-2
97%
MFCD07784376
290°C (dec.)
Videos
SDS