missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium 2-chlorophenyltrifluoroborate, 95%

Brand:  Alfa Aesar™ H37855.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15477827

  • £188.50 / 5g


Estimated Shipment date
from Supplier 05-01-2018
Add to basket

Specifications

Chemical Identifiers

Potassium 2-chlorophenyltrifluoroborate
218.46
MFCD09993172

Specifications

Potassium 2-chlorophenyltrifluoroborate
95%
MFCD09993172
>300°C
Powder
218.46
White
5g
Videos
SDS