missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium 2,2'-bithiophene-5-trifluoroborate, 95%

Brand:  Alfa Aesar™ H37541.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15417797

  • £65.30 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

Potassium 2,2'-bithiophene-5-trifluoroborate
272.17
MFCD26142849

Specifications

Potassium 2,2'-bithiophene-5-trifluoroborate
95%
MFCD26142849
Gray
1g
Powder
272.17
Air Sensitive
>300°C
Videos
SDS