missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potash, sulfurated, S 11.8% min as sulfide, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Potash is used in black/brown patina (copper, brass, bronze). Its use include decorative color effects on copper, brass, bronze and nickel. Sulfurated potash has been used in the treatment of acne and other skin disorders usually in the form of a lotion with zinc sulfate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Green |
Physical Form | Chunks |
Quantity | 100 g |
Assay Percent Range | S 11.8% min as sulfide |
Solubility Information | Soluble in water |
Sensitivity | Air sensitive; Hygroscopic |
Appearance | Green chunks |
Chemical Name or Material | Potash, Mixture composed of potassium polysulfides and potassium thiosulfate |
Safety and Handling
Product Identifier
- Potash, sulfated
- Danger
- Flammable solid Category 1
- Skin corrosion/irritation Category 1 B
- H228-Flammable solid.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'