Learn More
POT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566996
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: LTFEGTLGAPI IPRTSSKYFN FTTEDHKMVE ALRVWASTHM SPSWTLLKLC DVQPMQYFDL TCQLLGKAEV DGASFLLKVW DGTR Highest antigen sequence identity to the following orthologs - mouse 76%, rat 74%.
POT1 Polyclonal antibody specifically detects POT1 in Human samples. It is validated for Immunocytochemistry.Specifications
POT1 | |
Polyclonal | |
Unconjugated | |
POT1 | |
1500031H18Rik; AI505244; AI851169; CMM10; DKFZp586D211; Dolichyl-phosphate-mannose--protein mannosyltransferase 1; GLM9; HPOT1; mPot1; Pomt1; POT 1; POT1; POT-1; Pot1a; POT1-like telomere end-binding protein; protection of telomeres 1; protection of telomeres 1 homolog; protection of telomeres 1A; protection of telomeres protein 1; protein O-mannosyltransferase 1; Protein O-mannosyl-transferase 1; protein-O-mannosyltransferase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25913 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NUX5 | |
POT1 | |
Recombinant Human POT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only