Learn More
Brand™ Positive Displacement Tips
Manufactured from high quality polypropylene (cylinder) and HDPE (piston.) BRAND™ Positive Displacement Tips are suitable for dispensing fluids of high viscosity or high vapor pressure. Ideal for the BRAND™ repetitive pipet HandyStep™ electronic (with automatic tip size recognition) and HandyStep™ / HandyStep™ S. Individually wrapped to preserve sterility.
Brand:  Brand™ 702404
Code : 03
Additional Details : Weight : 0.32000kg
 View more versions of this product
Product Code. 10609472
  • £210.00 / Pack of 100
Estimated Shipment
26-06-2018
Add to basket
Description
Description
  • Manufactured from high quality polypropylene (cylinder) and HDPE (piston) *0.1mL version is made with LCP)
  • Suited for dispensing of fluids with high viscosity, high vapor pressure.
  • Ideal system component for the BRAND™ repetitive pipet HandyStep™ electronic (with automatic tip size recognition) and HandyStep™ / HandyStep™ S
  • Compliant with ISO 8655
  • Supplied with batch certificate and marked with the CE symbol according to the IVD Directive 98/79 EC.
  • Sterile and individually wrapped.
Specifications
Specifications
Individually Wrapped
Universal Fit Repeater Pipettor
Positive Displacement
Sterile
LCP/PP (Polypropylene)
HandyStep™ Electronic, HandyStep™ S, Universal Fit Repeater Pipettor
100 Pack
Individually Wrapped
0.1mL
No
ISO 8655, IVD Directive 98/79 EC
Videos
Documentation