missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Tall Form Melting Crucibles

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Tall Form Melting Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 79C5

UNSPSC : 41120000

Code : 02

Additional Details : Weight : 0.09400kg

 View more versions of this product

Product Code. 10332601

  • £10.30 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

120mL
60mm
Porcelain
60mm
75mm
Melting Crucible