missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Tall Form Crucibles

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Tall Form Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 79C/5

Code : 02

Additional Details : Weight : 0.09400kg

 View more versions of this product

Product Code. 12075083

  • £12.35 / Each


Estimated Shipment
18-06-2018
Add to basket

Specifications

Specifications

140mL
79mm
Porcelain
60mm
Melting Crucible
Videos