missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Tall Form Crucibles

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Tall Form Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 79C/0

Code : 02

Additional Details : Weight : 0.13536kg

 View more versions of this product

Product Code. 12386507

  • £68.09 / Pack of 10


In Stock 10
Add to basket

You may only order up to the available stock. For more details call customer services on 01509 555 500

Specifications

Specifications

15mL
38mm
Porcelain
30mm
Crucible
Videos
Documentation View accessory products