missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Spouted Mortar

Use these mortars for a variety of laboratory applications. Haldenwanger™ Porcelain Spouted Mortars are constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 55/15

Code : 02

Additional Details : Weight : 12.45000kg

 View more versions of this product

Product Code. 12393328

  • £712.24 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Grinding surfaces are unglazed.
Specifications

Specifications

6500mL
Mortar
140mm
Mortar
330mm
Porcelain
With spout
Videos
Documentation