missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Spouted Mortar

Brand:  Haldenwanger™ 55/8/rauh

UNSPSC : 41101702

Code : 02

Additional Details : Weight : 1.00000kg

Product Code. 10599981

  • £100.45 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

1000mL
Mortar
With spout
180mm
Porcelain
Mortar