missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Round Bottom Evaporating Basins

Brand:  Haldenwanger™ 109/10

UNSPSC : 41120000

Code : 02

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12025043

  • £223.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

Porcelain
310mm
310mm
Evaporating Basin
3400mL
110mm