missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Pestle

Use these pestles for a variety of laboratory applications. Haldenwanger™ Porcelain Pestles are constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 56/5

Code : 02

Additional Details : Weight : 0.22748kg

 View more versions of this product

Product Code. 12363338

  • £37.12 / Each


In Stock 4
Add to basket

Description

Description

  • Available in a variety of sizes
  • Grinding surfaces are unglazed
  • Complies with DIN VDE 0335, type C 100
Specifications

Specifications

48mm
Porcelain
Pestle
Pestle
185mm
Videos
Documentation View accessory products