missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Medium Form Melting Crucibles

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Medium Form Melting Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 79MF5

Code : 02

Additional Details : Weight : 0.03700kg

 View more versions of this product

Product Code. 10129490

  • £6.01 / Each


Estimated Shipment
14-02-2018
Add to basket

Specifications

Specifications

60mL
Glazed Inside and Outside Except Base
Porcelain
52mm
Melting Crucible
Videos