missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Medium Form Melting Crucibles

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Medium Form Melting Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 79MF/1A

UNSPSC : 41120000

Code : 02

Additional Details : Weight : 0.17000kg

 View more versions of this product

Product Code. 12095093

  • £23.55 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

300mL
89mm
75mm
Melting Crucible
89mm
Glaze
Porcelain