missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Medium Form Melting Crucibles

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Medium Form Melting Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 79MF/4

Code : 02

Additional Details : Weight : 0.04800kg

 View more versions of this product

Product Code. 12055093

  • £7.35 / Each


Estimated Shipment
20-06-2018
Add to basket

Specifications

Specifications

90mL
Glazed
Melting Crucible
60mm
50mm
Porcelain
Videos