missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Medium Form Melting Crucibles

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Medium Form Melting Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 79MF/6A

Code : 02

Additional Details : Weight : 0.03200kg

 View more versions of this product

Product Code. 12035093

  • £5.66 / Each


Estimated Shipment
12-06-2018
Add to basket

Specifications

Specifications

50mL
Glazed
Melting Crucible
50mm
40mm
Porcelain
Videos