missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Low Form Melting Crucibles

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Low Form Melting Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 79/4

Code : 02

Additional Details : Weight : 0.10700kg

 View more versions of this product

Product Code. 12005083

  • £12.70 / Each


Estimated Shipment
27-03-2018
Add to basket

Specifications

Specifications

180mL
Glazed
Melting Crucible
83mm
55mm
Porcelain
Videos