missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Low Form Melting Crucibles

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Low Form Melting Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 793

Code : 02

Additional Details : Weight : 2.63764kg

 View more versions of this product

Product Code. 10302791

  • £9.75 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

95mL
47mm
Porcelain
70mm
Melting Crucible
Videos