missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Low Form Melting Crucibles

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Low Form Melting Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 79/2a

Code : 02

Additional Details : Weight : 0.03700kg

 View more versions of this product

Product Code. 12081882

  • £7.66 / Each


Estimated Shipment
01-03-2018
Add to basket

Specifications

Specifications

65mL
38mm
Porcelain
60mm
Melting Crucible
Videos