missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Incinerating Dishes

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Incinerating Dishes feature a smooth internal finish, constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 33D4

Code : 02

Additional Details : Weight : 0.01500kg

 View more versions of this product

Product Code. 10662761

  • £7.27 / Each


Estimated Shipment
30-07-2018
Add to basket

Specifications

Specifications

Flat
47mm
Porcelain
10mL
12mm
Incinerating Dish
Videos