missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Incinerating Dishes

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Incinerating Dishes feature a smooth internal finish, constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 331

UNSPSC : 41121812

Code : 02

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 10587911

  • £8.69 / Each


In Stock 1
Add to basket

Specifications Description & Specifications

Specifications

20mL
20mm
Incinerating Dish
40mm
Porcelain