missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Hirsch Funnel

Brand:  Haldenwanger™ 126/4/X0A

Code : 02

Additional Details : Weight : 0.03500kg

 View more versions of this product

Product Code. 10538331

  • £25.84 / Each


Estimated Shipment
15-06-2018
Add to basket

Specifications

Specifications

13mm
With filter plate
1Each
Porcelain
13mm
Hirsch Funnel
61mm
Videos