missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Hirsch Funnel

Brand:  Haldenwanger™ 126/00

Code : 02

Additional Details : Weight : 0.08000kg

 View more versions of this product

Product Code. 12035123

  • £38.51 / Each


Estimated Shipment
19-12-2017
Add to basket

Specifications

Specifications

27mm
Hirsch Funnel
102mm
27mm
1Each
Porcelain
Videos