missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Hirsch Funnel

Brand:  Haldenwanger™ 126/000

Code : 02

Additional Details : Weight : 0.04500kg

 View more versions of this product

Product Code. 12025123

  • £30.16 / Each


Estimated Shipment
09-02-2018
Add to basket

Specifications

Specifications

15mm
Hirsch Funnel
74mm
15mm
1Each
Porcelain
Videos