Learn More
Porcelaines Avignon™ High Form Porcelain Crucibles
Heat and cool substances in the laboratory using Porcelaines Avignon™ Porcelain High Form Crucibles. They are available in various capacities.
£35.26
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11798473
|
Porcelaines Avignon™
11204 |
17 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- High walls
- Withstand high temperatures and rapid temperature changes
- Porcelain
Specifications
Porcelain | |
Crucible |