missing translation for 'onlineSavingsMsg'
Learn More

Porcelaines Avignon™ High Form Porcelain Crucibles

Heat and cool substances in the laboratory using Porcelaines Avignon™ Porcelain High Form Crucibles. They are available in various capacities.

Brand:  Porcelaines Avignon™ 11204

Code : A0

Additional Details : Weight : 0.11000kg

 View more versions of this product

Product Code. 11798473

  • £30.35 / Pack of 6


Estimated Shipment
08-03-2018
Add to basket

Description

Description

  • High walls
  • Withstand high temperatures and rapid temperature changes
  • Porcelain
Specifications

Specifications

17mL
40mm
Porcelain
35mm
Crucible
Videos